siRNA Details
| Gene Symbol: LINC00649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038883.2 | 2 | 650 | |||
| NR_134558.1 | 2 | 83 | ||||
| GenBank mRNA | AK311545.1 | - | 2 | 650 | ||
| AL532366.3 | - | 2 | 135 | |||
| CD699867.1 | - | 2 | 118 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00649| Gene Symbol: LINC00649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038883.2 | 2 | 650 | |||
| NR_134558.1 | 2 | 83 | ||||
| GenBank mRNA | AK311545.1 | - | 2 | 650 | ||
| AL532366.3 | - | 2 | 135 | |||
| CD699867.1 | - | 2 | 118 | |||