siRNA Details
| Gene Symbol: CCDC188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005261238.3 | XP_005261295.1 | 2 | 1373 | ||
| XM_005261239.3 | XP_005261296.1 | 2 | 1374 | |||
| XM_005261240.3 | XP_005261297.1 | 2 | 1372 | |||
| XM_011530170.2 | XP_011528472.1 | 2 | 1373 | |||
| XM_011530171.2 | XP_011528473.1 | 2 | 1373 | |||

CCDC188
Genomic Map