siRNA Details
| Gene Symbol: FAM227A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013647.1 | NP_001013669.1 | 17 | 2385 | ||
| NM_001291030.1 | NP_001277959.1 | 17 | 2425 | |||
| XM_006724293.2 | XP_006724356.1 | 17 | 2397 | |||
| GenBank mRNA | AK301819.1 | - | 17 | 2099 | ||

FAM227A
Genomic Map