siRNA Details
| Gene Symbol: DNAJC27-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034113.1 | 1 | 52 | |||
| GenBank mRNA | CR933670.1 | - | 1 | 37 | ||
| DA314471.1 | - | 1 | 52 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC27-AS1| Gene Symbol: DNAJC27-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034113.1 | 1 | 52 | |||
| GenBank mRNA | CR933670.1 | - | 1 | 37 | ||
| DA314471.1 | - | 1 | 52 | |||