siRNA Details
| Gene Symbol: IL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018724.3 | NP_061194.2 | 3 | 330 | ||
| XM_005273152.3 | XP_005273209.1 | 4 | 756 | |||
| GenBank mRNA | AF224266.1 | - | 3 | 330 | ||
| AY358320.1 | - | 4 | 649 | |||
| AY876922.1 | - | 3 | 286 | |||
| BC069311.1 | - | 3 | 286 | 7262116 | ||
| BC069364.1 | - | 3 | 286 | 7262128 | ||
| BC069398.1 | - | 3 | 286 | 7262140 | ||
| BC069425.1 | - | 3 | 286 | 7262152 | ||
| BC069449.1 | - | 3 | 286 | 7262164 | ||
| BC069487.1 | - | 3 | 286 | 7262176 | ||
| BC069523.1 | - | 3 | 286 | 7262093 | ||
| BC069559.1 | - | 3 | 286 | 7262105 | ||
| BC074948.2 | - | 3 | 311 | 30915498 | ||
| BC074949.2 | - | 3 | 311 | 30915275 | ||
| BC117386.1 | - | 3 | 312 | 40125937 | ||
| BC117390.1 | - | 3 | 312 | 40125941 | ||

IL20
Genomic Map