siRNA Details
| Gene Symbol: DPP10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036580.1 | 3 | 430 | |||
| GenBank mRNA | AB593078.1 | - | 3 | 452 | ||
| BC032913.2 | - | 3 | 430 | |||
| BC048425.1 | - | 3 | 423 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPP10-AS1| Gene Symbol: DPP10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036580.1 | 3 | 430 | |||
| GenBank mRNA | AB593078.1 | - | 3 | 452 | ||
| BC032913.2 | - | 3 | 430 | |||
| BC048425.1 | - | 3 | 423 | |||