siRNA Details
| Gene Symbol: LOC100507460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648270.1 | - | 4 | 1923 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507460| Gene Symbol: LOC100507460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX648270.1 | - | 4 | 1923 | ||