siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290030.1 | NP_001276959.1 | 1 | 14 | ||
| NM_001290031.1 | NP_001276960.1 | 1 | 14 | |||
| NR_110644.1 | 1 | 14 | ||||
| XM_011510998.2 | XP_011509300.1 | 2 | 488 | |||
| XM_011510999.2 | XP_011509301.1 | 1 | 155 | |||
| XM_017003875.1 | XP_016859364.1 | 2 | 515 | |||
| XM_017003877.1 | XP_016859366.1 | 1 | 155 | |||

ERICH2
Genomic Map