siRNA Details
| Gene Symbol: RBMS3-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109804.1 | 1 | 183 | |||
| GenBank mRNA | BM840572.1 | - | 1 | 183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMS3-AS3| Gene Symbol: RBMS3-AS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109804.1 | 1 | 183 | |||
| GenBank mRNA | BM840572.1 | - | 1 | 183 | ||