siRNA Details
| Gene Symbol: IQCF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145059.1 | NP_001138531.1 | 2 | 104 | ||
| GenBank mRNA | BX106781.1 | - | 2 | 104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IQCF5| Gene Symbol: IQCF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145059.1 | NP_001138531.1 | 2 | 104 | ||
| GenBank mRNA | BX106781.1 | - | 2 | 104 | ||