siRNA Details
| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110167.1 | 3 | 326 | |||
| NR_110168.1 | 3 | 279 | ||||
| GenBank mRNA | BF676681.1 | - | 3 | 328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01213| Gene Symbol: LINC01213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110167.1 | 3 | 326 | |||
| NR_110168.1 | 3 | 279 | ||||
| GenBank mRNA | BF676681.1 | - | 3 | 328 | ||