siRNA Details
| Gene Symbol: TMEM212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164436.1 | NP_001157908.1 | 5 | 1255 | ||
| GenBank mRNA | AK026825.1 | - | 5 | 1235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM212| Gene Symbol: TMEM212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164436.1 | NP_001157908.1 | 5 | 1255 | ||
| GenBank mRNA | AK026825.1 | - | 5 | 1235 | ||