siRNA Details
| Gene Symbol: LOC728339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC087857.1 | - | 3 | 670 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC728339| Gene Symbol: LOC728339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC087857.1 | - | 3 | 670 | ||