siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098176.1 | - | 16 | 2132 | ||
| AK302348.1 | - | 14 | 1633 | |||
| AK304443.1 | - | 16 | 1794 | |||
| AK316511.1 | - | 16 | 2228 | |||
| Gene Symbol: LOC107984125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014157.1 | XP_016869646.1 | 9 | 1341 | ||
| XM_017014158.1 | XP_016869647.1 | 4 | 677 | |||

RP1
Genomic Map