siRNA Details
| Gene Symbol: CNTNAP3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK024257.1 | - | 9 | 2576 | ||
| Gene Symbol: LOC100289279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011545673.2 | XP_011543975.1 | 8 | 2583 | ||

CNTNAP3P2
Genomic Map