siRNA Details
| Gene Symbol: CNTNAP3P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_111893.2 | 24 | 5435 | |||
| Gene Symbol: CNTNAP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201380.2 | NP_001188309.2 | 24 | 5746 | ||
| GenBank mRNA | BX538190.1 | - | 5 | 2014 | ||

CNTNAP3P2
Genomic Map