siRNA Details
| Gene Symbol: NUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138459.3 | NP_612468.1 | 5 | 3198 | ||
| GenBank mRNA | AJ420576.1 | - | 1 | 1467 | ||
| AK025467.1 | - | 1 | 1429 | |||
| AK125119.1 | - | 5 | 3160 | |||
| AK126335.1 | - | 6 | 2911 | |||
| BC110325.1 | - | 5 | 3153 | 5553827 | ||

NUS1
Genomic Map