siRNA Details
| Gene Symbol: LOC105373381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011531226.2 | XP_011529528.1 | 1, 2 | 2170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105373381| Gene Symbol: LOC105373381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011531226.2 | XP_011529528.1 | 1, 2 | 2170 | ||