siRNA Details
| Gene Symbol: GUCY2GP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028134.1 | 16 | 2622 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY2GP| Gene Symbol: GUCY2GP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028134.1 | 16 | 2622 | |||