siRNA Details
| Gene Symbol: GUCY2EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024042.1 | 12 | 1457 | |||
| GenBank mRNA | AB231718.1 | - | 1 | 20 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY2EP| Gene Symbol: GUCY2EP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024042.1 | 12 | 1457 | |||
| GenBank mRNA | AB231718.1 | - | 1 | 20 | ||