siRNA Details
| Gene Symbol: CLN8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018941.3 | NP_061764.2 | 3 | 2229 | ||
| GenBank mRNA | AF123758.1 | - | 3 | 2159 | ||
| AF123759.1 | - | 3 | 2159 | |||
| AF123760.1 | - | 3 | 2159 | |||
| AF123761.1 | - | 3 | 2159 | |||

CLN8
Genomic Map