siRNA Details
| Gene Symbol: ONECUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080488.1 | NP_001073957.1 | 2 | 1297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ONECUT3| Gene Symbol: ONECUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080488.1 | NP_001073957.1 | 2 | 1297 | ||