siRNA Details
| Gene Symbol: SBK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199824.2 | NP_001186753.1 | 3 | 253 | ||
| XM_011526298.2 | XP_011524600.1 | 3 | 344 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SBK3| Gene Symbol: SBK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199824.2 | NP_001186753.1 | 3 | 253 | ||
| XM_011526298.2 | XP_011524600.1 | 3 | 344 | |||