siRNA Details
| Gene Symbol: GNG12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040077.1 | 1 | 21 | |||
| GenBank mRNA | DB291468.1 | - | 1 | 23 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNG12-AS1| Gene Symbol: GNG12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040077.1 | 1 | 21 | |||
| GenBank mRNA | DB291468.1 | - | 1 | 23 | ||