siRNA Details
| Gene Symbol: SNTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018968.3 | NP_061841.2 | 16 | 1550 | ||
| XM_011510366.2 | XP_011508668.1 | 14 | 1587 | |||
| XM_017004361.1 | XP_016859850.1 | 16 | 1512 | |||
| XM_017004362.1 | XP_016859851.1 | 17 | 1751 | |||
| XM_017004363.1 | XP_016859852.1 | 16 | 1699 | |||
| XM_017004364.1 | XP_016859853.1 | 13 | 4840 | |||
| XM_017004367.1 | XP_016859856.1 | 17 | 1588 | |||
| XM_017004373.1 | XP_016859862.1 | 11 | 2154 | |||
| GenBank mRNA | AJ003029.1 | - | 16 | 1552 | ||
| BC125251.1 | - | 10 | 1098 | 40025232 | ||
| BG208326.1 | - | 2 | 136 | |||

SNTG2
Genomic Map