siRNA Details
| Gene Symbol: YIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018982.4 | NP_061855.1 | 4 | 521 | ||
| NR_036639.1 | 4 | 521 | ||||
| NR_036640.1 | 3 | 301 | ||||
| NR_135075.1 | 2 | 238 | ||||
| XM_017001499.1 | XP_016856988.1 | 3 | 279 | |||
| GenBank mRNA | AK000823.1 | - | 2 | 186 | ||
| AK300240.1 | - | 3 | 383 | |||
| AK315183.1 | - | 4 | 499 | |||
| AL713668.1 | - | 3 | 232 | |||
| BC064590.1 | - | 4 | 487 | 5749036 | ||
| BI561906.1 | - | 4 | 548 | |||
| BI757212.1 | - | 4 | 518 | |||
| BX405963.2 | - | 4 | 513 | |||
| CN308348.1 | - | 4 | 522 | |||

YIPF1
Genomic Map