siRNA Details
| Gene Symbol: USP17L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256874.1 | NP_001243803.1 | 1 | 1472 | ||
| Gene Symbol: USP17L8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256872.1 | NP_001243801.1 | 1 | 1472 | ||

USP17L4
Genomic Map