siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098176.1 | - | 6 | 779 | ||
| AK304443.1 | - | 5 | 540 | |||
| AK316511.1 | - | 5 | 974 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RP1| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098176.1 | - | 6 | 779 | ||
| AK304443.1 | - | 5 | 540 | |||
| AK316511.1 | - | 5 | 974 | |||