siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098176.1 | - | 7 | 913 | ||
| AK302348.1 | - | 4 | 513 | |||
| AK304443.1 | - | 6 | 674 | |||
| AK316511.1 | - | 6 | 1108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RP1| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098176.1 | - | 7 | 913 | ||
| AK302348.1 | - | 4 | 513 | |||
| AK304443.1 | - | 6 | 674 | |||
| AK316511.1 | - | 6 | 1108 | |||