siRNA Details
| Gene Symbol: TMEM55A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018710.2 | NP_061180.1 | 7 | 1700 | ||
| GenBank mRNA | AX775961.1 | - | 7 | 1549 | ||
| BC033892.1 | - | 7 | 1549 | 4820065 | ||
| CR749733.1 | - | 6 | 1263 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM55A| Gene Symbol: TMEM55A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018710.2 | NP_061180.1 | 7 | 1700 | ||
| GenBank mRNA | AX775961.1 | - | 7 | 1549 | ||
| BC033892.1 | - | 7 | 1549 | 4820065 | ||
| CR749733.1 | - | 6 | 1263 | |||