siRNA Details
| Gene Symbol: CDCA7L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127370.2 | NP_001120842.1 | 5 | 583 | ||
| NM_001127371.2 | NP_001120843.1 | 3 | 391 | |||
| NM_018719.4 | NP_061189.2 | 4 | 529 | |||
| GenBank mRNA | AK022955.1 | - | 4 | 504 | ||
| AK095965.1 | - | 5 | 537 | |||
| AK292490.1 | - | 4 | 504 | |||
| AK300781.1 | - | 3 | 351 | |||
| AY161168.1 | - | 4 | 406 | |||
| AY161169.1 | - | 4 | 409 | |||
| BC009352.2 | - | 4 | 421 | 4139111 | ||
| BC014630.2 | - | 3 | 373 | |||
| BC025242.1 | - | 4 | 483 | 4842169 | ||
| BC032576.1 | - | 4 | 494 | 5491780 | ||
| BC039823.1 | - | 4 | 493 | 6057601 | ||
| BX422040.2 | - | 3 | 339 | |||
| DA041070.1 | - | 4 | 529 | |||
| DC331792.1 | - | 3 | 391 | |||

CDCA7L
Genomic Map