siRNA Details
| Gene Symbol: PCNX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032223.3 | NP_115599.2 | 26 | 4025 | ||
| XM_005273987.4 | XP_005274044.1 | 26 | 5945 | |||
| XM_006718549.3 | XP_006718612.1 | 26 | 5930 | |||
| XM_006718551.3 | XP_006718614.1 | 26 | 5948 | |||
| XM_011545024.2 | XP_011543326.1 | 27 | 5121 | |||
| XM_011545025.2 | XP_011543327.1 | 26 | 5948 | |||
| XM_011545026.2 | XP_011543328.1 | 20 | 2788 | |||
| XM_011545027.1 | XP_011543329.1 | 18 | 2001 | |||
| GenBank mRNA | BX640978.1 | - | 6 | 697 | ||
| CA489689.1 | - | 3 | 214 | |||

PCNX3
Genomic Map