siRNA Details
| Gene Symbol: LINC00942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028415.1 | 2 | 409 | |||
| GenBank mRNA | AK090616.1 | - | 2 | 373 | ||
| BG332174.1 | - | 409 | ||||
| BQ942184.1 | - | 2 | 315 | |||
| BQ954540.1 | - | 2 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00942| Gene Symbol: LINC00942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028415.1 | 2 | 409 | |||
| GenBank mRNA | AK090616.1 | - | 2 | 373 | ||
| BG332174.1 | - | 409 | ||||
| BQ942184.1 | - | 2 | 315 | |||
| BQ954540.1 | - | 2 | 330 | |||