siRNA Details
| Gene Symbol: LINC00939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034132.1 | 3 | 1198 | |||
| GenBank mRNA | AK057632.1 | - | 4 | 911 | ||
| AL137270.1 | - | 4 | 990 | |||
| BC057846.1 | - | 3 | 1196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00939| Gene Symbol: LINC00939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034132.1 | 3 | 1198 | |||
| GenBank mRNA | AK057632.1 | - | 4 | 911 | ||
| AL137270.1 | - | 4 | 990 | |||
| BC057846.1 | - | 3 | 1196 | |||