siRNA Details
| Gene Symbol: FLJ22447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039985.1 | 2 | 257 | |||
| GenBank mRNA | AK026100.1 | - | 2 | 261 | ||
| BQ439352.1 | - | 2 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ22447| Gene Symbol: FLJ22447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039985.1 | 2 | 257 | |||
| GenBank mRNA | AK026100.1 | - | 2 | 261 | ||
| BQ439352.1 | - | 2 | 346 | |||