siRNA Details
| Gene Symbol: LOC643802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207030.1 | NP_001193959.1 | 2 | 1094 | ||
| XM_017023560.1 | XP_016879049.1 | 3 | 679 | |||
| GenBank mRNA | BX537874.1 | - | 2 | 935 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643802| Gene Symbol: LOC643802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207030.1 | NP_001193959.1 | 2 | 1094 | ||
| XM_017023560.1 | XP_016879049.1 | 3 | 679 | |||
| GenBank mRNA | BX537874.1 | - | 2 | 935 | ||