siRNA Details
| Gene Symbol: LOC400541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096066.1 | - | 2, 3 | 779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC400541| Gene Symbol: LOC400541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK096066.1 | - | 2, 3 | 779 | ||