siRNA Details
| Gene Symbol: LINC00675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036581.1 | 2 | 518 | |||
| GenBank mRNA | BC015790.1 | - | 2 | 456 | ||
| BC041634.1 | - | 2 | 456 | |||
| BC063383.1 | - | 2 | 91 | |||
| BM924455.1 | - | 2 | 471 | |||
| DA425830.1 | - | 2 | 518 | |||

LINC00675
Genomic Map