siRNA Details
| Gene Symbol: PCDHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018905.2 | NP_061728.1 | 1 | 735 | ||
| NM_031495.1 | NP_113683.1 | 1 | 735 | |||
| NM_031496.1 | NP_113684.1 | 1 | 735 | |||
| GenBank mRNA | AF152310.1 | - | 1 | 629 | ||
| AF152480.1 | - | 1 | 629 | |||
| BC003126.1 | - | 1 | 735 | 3503666 | ||

PCDHA2
Genomic Map