siRNA Details
| Gene Symbol: LINC00668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034100.1 | 4 | 647 | |||
| GenBank mRNA | AK095347.1 | - | 3 | 558 | ||
| BC035413.1 | - | 4 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00668| Gene Symbol: LINC00668 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034100.1 | 4 | 647 | |||
| GenBank mRNA | AK095347.1 | - | 3 | 558 | ||
| BC035413.1 | - | 4 | 647 | |||