siRNA Details
| Gene Symbol: SLC14A2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110899.1 | 5 | 685 | |||
| GenBank mRNA | AK126075.1 | - | 5 | 685 | ||
| BX109976.1 | - | 11 | ||||
| HY036726.1 | - | 1 | 54 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC14A2-AS1| Gene Symbol: SLC14A2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110899.1 | 5 | 685 | |||
| GenBank mRNA | AK126075.1 | - | 5 | 685 | ||
| BX109976.1 | - | 11 | ||||
| HY036726.1 | - | 1 | 54 | |||