siRNA Details
| Gene Symbol: LINC01630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040075.1 | 2 | 844 | |||
| GenBank mRNA | AK094936.1 | - | 2 | 844 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01630| Gene Symbol: LINC01630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040075.1 | 2 | 844 | |||
| GenBank mRNA | AK094936.1 | - | 2 | 844 | ||