siRNA Details
| Gene Symbol: LINC00683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120419.1 | 2 | 1744 | |||
| GenBank mRNA | AK094957.1 | - | 2 | 652 | ||
| AK127565.1 | - | 2 | 1745 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00683| Gene Symbol: LINC00683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120419.1 | 2 | 1744 | |||
| GenBank mRNA | AK094957.1 | - | 2 | 652 | ||
| AK127565.1 | - | 2 | 1745 | |||