siRNA Details
Gene Symbol: LOC105376805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135058.1 | 1 | 99 | |||
NR_135059.1 | 1 | 99 | ||||
GenBank mRNA | BM923306.1 | - | 1 | 39 | ||
BX115769.1 | - | 1 | 74 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105376805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135058.1 | 1 | 99 | |||
NR_135059.1 | 1 | 99 | ||||
GenBank mRNA | BM923306.1 | - | 1 | 39 | ||
BX115769.1 | - | 1 | 74 |