siRNA Details
| Gene Symbol: FAM229A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167676.1 | NP_001161148.1 | 3 | 672 | ||
| GenBank mRNA | AK125095.1 | - | 2 | 2733 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001085491, AK018987, AK006760, AK018907, AK018964, and others.

FAM229A
Genomic Map