siRNA Details
| Gene Symbol: NOL4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256798.1 | NP_001243727.1 | 1 | 319 | ||
| XM_011528562.2 | XP_011526864.1 | 1 | 313 | |||
| XM_011528563.1 | XP_011526865.1 | 1 | 313 | |||
| GenBank mRNA | BC023657.2 | - | 1 | 319 | ||
| BC065483.1 | - | 1 | 319 | |||
| BQ067609.1 | - | 1 | 74 | |||

NOL4L
Genomic Map