siRNA Details
Gene Symbol: LOC729296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109916.1 | 5 | 1096 | |||
NR_109917.1 | 5 | 866 | ||||
NR_109918.1 | 5 | 763 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC729296 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109916.1 | 5 | 1096 | |||
NR_109917.1 | 5 | 866 | ||||
NR_109918.1 | 5 | 763 |