siRNA Details
Gene Symbol: LINC00163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033840.1 | 2 | 2012 | |||
GenBank mRNA | BC036902.1 | - | 2 | 2148 | ||
BC046564.1 | - | 2 | 2117 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033840.1 | 2 | 2012 | |||
GenBank mRNA | BC036902.1 | - | 2 | 2148 | ||
BC046564.1 | - | 2 | 2117 |