siRNA Details
| Gene Symbol: LINC01122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033873.1 | 14 | 1534 | |||
| GenBank mRNA | AK055400.1 | - | 14 | 1534 | ||
| AL832565.1 | - | 7 | 537 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01122| Gene Symbol: LINC01122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033873.1 | 14 | 1534 | |||
| GenBank mRNA | AK055400.1 | - | 14 | 1534 | ||
| AL832565.1 | - | 7 | 537 | |||