siRNA Details
| Gene Symbol: TEX41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033870.2 | 5 | 642 | |||
| GenBank mRNA | BC043549.1 | - | 5 | 583 | ||
| BX648102.1 | - | 4 | 487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEX41| Gene Symbol: TEX41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033870.2 | 5 | 642 | |||
| GenBank mRNA | BC043549.1 | - | 5 | 583 | ||
| BX648102.1 | - | 4 | 487 | |||